Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]

Supplier: Enzo Life Sciences
ADI-SPA-827-J
95046-234EA 2332.6 USD
95046-234
Anti-KDEL Mouse Monoclonal Antibody [clone: 10C3]
Antibodies
The tetrapeptide KDEL, located at the carboxy-terminal sequences of luminal proteins, is a retrieval motif essential for the precise sorting of these proteins along the secretory pathway. KDEL proteins perform essential functions in the endoplasmic reticulum (ER) related to protein folding as well as assembly. The localization of chaperones and other soluble proteins to the ER is achieved by their continuous retrieval from post-ER compartments by the KDEL receptor (Erd2p), which is a membrane protein localized in the Golgi apparatus.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, EM

Recognizes proteins containing the KDEL sequence in mammals, birds, insects, plants, and yeast. Detects three bands of ~94kDa, ~78 kDa and ~55kDa by Western blot..

Type: Primary
Antigen: KDEL
Clonality: Monoclonal
Clone: 10C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mammals
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR