SklarSafe™ Safety Scalpels, Non sterile, Sklar
Supplier: Sklar
06-3132
95045-498CS 1218.25 USD
95045-498
SklarSafe™ Safety Scalpels, Non sterile, Sklar
Scalpels and Scalpel Assemblies Scalpels
The SklarSafe™ Safety Scalpel features a protective, transparent sheath that can be opened, closed, and locked with a single hand
The sturdy "sure-grip" handle provides an audible click when the sheath is advanced to a locked position. This instrument is OSHA compliant and individually sealed in sturdy film and Tyvek packaging. Nonsterile, disposable scalpels are supplied in a box of 10.