Tween-20 solution 10%, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-80-1929
95045-198EA 82.84 USD
95045-198
Tween-20 solution 10%, Enzo Life Sciences
Buffers Assay Buffers
This Tween®-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol.
The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.