Glass Plates, Cytiva
Supplier: Cytiva
80-6190-01
95044-744PK 281.93 USD
95044-744 95040-386
Glass Plates, Cytiva
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Polyacrylamide IEF gels can be cast in a cassette system.
The cassette is made up by a glass plate with the film support and an U-frame, and is hold together with clamps.