Hybridization Solution I, G-Biosciences
Supplier: G-Biosciences
786-528
95043-518EA 107.34 USD
95043-518
Hybridization Solution I, G-Biosciences
Buffers Electrophoresis Buffers Hybridization Buffers
G-Biosciences' Hybridization Solution I contains: 0.9 M NaCl and 90 mM Trisodium citrate, with 5% Tween® 20.
Ordering information: Accessories available.