Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Supplier: G-Biosciences
786-499
95043-428EA 107.34 USD
95043-428
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Electrophoresis Reagents Stain Removal Reagents
After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution.
  • Contains Methanol and Acetic acid