Hematoxylin modified solution (Harris)
Supplier: VWR International
Synonyms:
DcH-48, Natural Black I, Hydroxybrasilin
ES702
95043-368CS
1062.26
USD
95043-368
Hematoxylin modified solution (Harris)
Hematoxylin staining solutions
This regressive nuclear stain is much like Mayer’s Hematoxylin but is more stable.
- No Mercury
- Ready to Use Formula
- Exceptional Nuclear Detail
The formulation does not contain mercury.
This solution is ready to use and provides exceptional nuclear detail when used in progressive or regressive protocols.
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...