Surgical Instrument Cleaner and Lubricant, Integra™ Miltex®
Supplier: Miltex
3-710
95042-720EA 87.78 USD
95042-720
Surgical Instrument Cleaner and Lubricant, Integra™ Miltex®
Cleaning Agents Detergents and Surface Cleansers
Dual-purpose instrument cleaner and lubricant is chemically engineered for use as a presoak or for manual cleaning in washers, decontaminators, ultrasonic cleaners, and cart washers
The biodegradable solution ensures cleaner and brighter instruments, and will remove stains, blood, and mineral deposits.