Surgical Instrument Stain Remover, Integra™ Miltex®
Supplier: Miltex
3-740
95042-716CS 238.36 USD
95042-716
Surgical Instrument Stain Remover, Integra™ Miltex®
Cleaning Agents Detergents and Surface Cleansers
Powdered formula removes stains and tarnish, and restores luster to surgical instruments, autoclaves, ultrasonic cleaners, stainless steel countertops, tiles, and plated surfaces
Effective on a variety of materials including stainless steel, chrome, nickel, brass, copper, and silver. The formula will not harm instrument surfaces.
Ordering information: Stain remover is packaged in an 85g (3oz.) jar. Jars are supplied in cases of 12.