VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
95042-566
95042-566PK 117.07 USD
95042-566
VWR® Chemical-Resistant Laboratory Marker
Pens Lab Markers
Permanently marks slides and cassettes in the histology laboratory.
  • Ultra-fine tip
  • Ink is waterproof, smearproof, and odorless
  • Resistant to common histology chemicals, including formalin, xylene, and alcohols
Note: Markers are not for use on smooth surfaces; only on paint frosted areas of slides and cassettes.