VWR® Pour Spout Sample Cups
Supplier: VWR International
95042-200
95042-200CS 237.46 USD
95042-200
VWR® Pour Spout Sample Cups
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
These translucent polystyrene containers are appropriate for use as urine cups or many other applications.
  • Polystyrene construction
  • Translucent siding
  • Suitable for a range of applications
They are graduated in ounces, cc, and mililiters, and have convenient pour spouts. They are packaged in sleeves of 25.