Sterile New Sponge Dressings, DUKAL™ Corporation
Supplier: Dukal
6122 6123 6124 6125
95041-752CS 238.45 USD
95041-752 95041-756 95041-760 95041-764
Sterile New Sponge Dressings, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Sterile New Sponges are a rayon/poly blend construction that are virtually non-linting.
  • The effective combination of weight and open aperture allows each sponge to wick quicker and offer superior absorbency
  • Virtually non-linting
  • Not made with natural rubber latex