Xylene (mixture of isomers), Lab-Grade for histology
Supplier: VWR International
|
Danger
|
Laboratory grade quality suitable for procedures in Histology
- For clearing, deparaffinizing
Caution: This product can expose you to chemicals including Ethylbenzene, which is known to the State of California to cause cancer.
Formula:
C₆H₄(CH₃)₂ Boiling Pt: 137…140℃ |
MDL Number:
MFCD00077264 CAS Number: 1330-20-7 UN: 1307 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...