2-Propanol ≥99%, purified for histology
Supplier: VWR International
|
Danger
|
95041-468
95041-468CS
546.26
USD
95041-468
2-Propanol ≥99%, purified for histology
2-Propanol
99% Isopropyl Alcohol, Purified
- Ideal for specialty fixation and dehydration in the histology lab
Formula:
(CH₃)₂CHOH |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...