Methanol for histology

Supplier: VWR International
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

ES607
95041-464CS 203.54 USD
95041-464
Methanol for histology
Methanol

Methyl Alcohol, anhydrous


  • Used for fixation of blood and bone marrow smears and cytology specimens
  • Ideal for specialty fixation and dehydration in the histology lab


Caution: This product can expose you to chemicals including Methanol, which is known to the State of California to cause birth defects or other reproductive harm

Formula: CH₃OH
MW: 32.04 g/mol
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR