Alcohol 100%, Reagent Grade for histology (RGA 100)

Supplier: VWR International
Danger

95041-462
95041-462CS 168.65 USD
95041-462
Alcohol 100%, Reagent Grade for histology (RGA 100)
Alcohol

Anhydrous reagent grade alcohol is a denatured form of ethyl alcohol, approved for sale under U.S. regulations (90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol).


  • Used in histology tissue preparation


It is a preferred mixture for dehydrationg tissue samples prior to embedding in histology.


Caution: This product can expose you to chemicals including Methanol, which is known to the State of California to cause birth defects or other reproductive harm

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR