Anti-ERK1 + ERK2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-MA001-D
95040-468EA 241.98 USD
95040-468
Anti-ERK1 + ERK2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Drosophila, Chicken, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR