Microarray Hybridization Solution v2.0, Cytiva
Supplier: Cytiva
RPK0325
95040-374EA 215.94 USD
95040-374
Microarray Hybridization Solution v2.0, Cytiva
Microarray Reagents
Microarray Hybridization Solution Version 2.0, for highly reproducible microarray results.
  • Includes a blend of proprietary components to provide highly reproducible, high-quality automated and manual DNA microarray hybridizations
  • Hybridization rate enhancers improve the detection of rare targets
  • Proprietary blocking agents minimize non-specific background
DNA microarrays can be used to measure the expression levels of large numbers of genes simultaneously (gene expression profiling), or to genotype multiple areas of a genome (SNP arrays), with variable coverage or resolution depending how many DNA 'spots' cover those areas.