First-Strand cDNA Synthesis Kit, Cytiva
Supplier: Cytiva
27-9261-01
95040-330EA 693.57 USD
95040-330
First-Strand cDNA Synthesis Kit, Cytiva
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Designed to generate full length first-strand cDNA from mRNA templates. Following first-strand cDNA synthesis, the sample can be used directly for in vitro amplification using PCR or for second-strand synthesis using the Gubler-Hoffman method.
  • For synthesis of single-stranded cDNA prior to amplification by PCR
  • Features preassembled bulk reaction mixes to save time and minimise potential for pipetting errors
  • Allows users to choose primers
  • Includes a choice of two primers: pd(N)6 random hexamers and NotI-(dT)18
Delivery information: Kit includes sufficient for up to 55 first-strand syntheses, each of which will produce enough material for up to 15 amplification reactions: Bulk First-Strand cDNA Reaction Mixes, mRNA Standard, DTT Solution, RNase-Free Water, pd(N)6 Primer, NotI-(dT)18 Bifunctional Primer and instruction booklet.
Caution: Storage -15 to -30°C