Amersham™ CyDye™ Fluorescent Nucleotides
Supplier: Cytiva
PA53021 PA53521 PA55021 PA55322 PA53022 PA53026 PA55022 PA53031 PA55321
95040-140EA 599.43 USD
95040-140 95040-142 95040-144 95040-154 95040-156 95040-160 95040-168 95040-146 95040-148 95040-158 95040-150 95040-166
Amersham™ CyDye™ Fluorescent Nucleotides
Protein Linking and Labeling Systems
Fluorescent nucleotides for labeling DNA and RNA probes, especially during multi-colour analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
  • High sensitivity and low non specific binding
  • High photostability, high water solubility
  • pH insensitive
CyDye™ labelled nucleotides are available in a range of bright and intense colours with narrow emission bands for multiplexing with a single sample.