Deaza dGTP, Cytiva
Supplier: Cytiva
27-2090-02
95038-208EA 388.82 USD
95038-208
Deaza dGTP, Cytiva
Nucleic Acid Reagents Nucleotides
Band compressions are ambiguous band patterns on sequencing gels caused by stable intra-strand secondary structures that are not fully denatured during electrophoresis. Their occurrence can be reduced by substituting nucleotide analogues in the sequencing mixes. 7-Deaza dGTP (c⁷dGTP) can be used to resolve G-band compressions.
  • For reducing 'band compressions' in sequencing gels
  • Supplied as a 5 mM solution