D-ɑ-Tocopherol succinate ≥96.0% USP
Supplier: Spectrum Chemical Mfg. Corp.
RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₃₃H₅₄O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00072055 CAS Number: 4345-03-3 |
Specification Test Results
Assay | 96.0 - 102.0 % |
Acidity | To Pass Test |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...