Sorbic acid ≥99%, powder NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

SO215-2.5KG SO215-100GM SO215-500GM SO215-12KG
95036-004EA 611.15 USD
95036-004 95036-002 95036-006 700005-522
Sorbic acid ≥99%, powder NF suitable for use as excipient
Sorbic acid

Sorbic Acid, Powder, NF is used as an antimicrobial agent in pharmaceutical formulations. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228
Melting Pt: 132…135 °C
Density: 1.204 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR