Saccharin sodium dihydrate ≥99.0%, Powder USP

Supplier: Spectrum Chemical Mfg. Corp.

SO200-100GM SO200-2.5KG SO200-500GM SO200-12KG SO200-25GM
95035-924EA 226.8 USD
95035-924 95035-926 95035-928 75812-476 75812-478
Saccharin sodium dihydrate ≥99.0%, Powder USP
Saccharin sodium dihydrate

Saccharin Sodium, Dihydrate, Powder, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₇H₅NNaO₃S·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 101.0 %
Clarity of Solution To Pass Test
Color of Solution To Pass Test
Acidity or Alkalinity To Pass Test
Water Determination max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR