Sodium dodecyl sulfate (SDS) ≥85% BP, Ph. Eur., JP, NF suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 204…207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 2811 ADR: 6.1,III Merck Index: 14,08636 |
|
Specification Test Results
Assay | 85% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...