Sodium carbonate, anhydrous ≥99.5% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Soda, Carbonic acid sodium salt, Calcined soda, Disodium carbonate, Carbonic acid disodium salt, Na-X, Bisodium carbonate, Carbonic acid, disodium salt, Soda ash

SO147-12KG SO147-500GM SO147-2.5KG SO147-50KGBL
89050-460EA 1013.83 USD
89050-460 95035-744 95035-742 75812-434
Sodium carbonate, anhydrous ≥99.5% NF
Sodium carbonate

Sodium Carbonate, Anhydrous, NF is a pH regulator excipient. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Na₂CO₃) 99.5 - 100.5 %
Water max. 0.5 %
Heavy Metals (As Pb) max. 0.001 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR