Sodium benzoate ≥99%, powder NF suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Benzoic acid sodium salt
SO120-500GM
SO120-12KG
SO120-2.5KG
SO120-50KGBL
95035-712EA
216.42
USD
95035-712
700005-628
95035-710
89078-998
Sodium benzoate ≥99%, powder NF suitable for use as excipient
Sodium benzoate
Sodium Benzoate, Powder, NF is a tablet and capsule lubricant used in the pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₆H₅COONa Melting Pt: 410…430 °C Density: 1.44 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012463 CAS Number: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...