Riboflavine (vitamin B2) ≥98.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

RI103-50KGBL RI103-100GM RI103-1KG RI103-500GM RI103-12KGBL RI103-25GM RI103-25KGBL
700009-334EA 9365.1 USD
700009-334 95035-556 95035-558 95035-562 700009-332 95035-560 75812-116
Riboflavine (vitamin B2) ≥98.0% USP
Riboflavine (Vitamin B2)

Riboflavin, USP is also referred to as vitamin B2 and used as a dietary supplement.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₁₇H₂₀N₄O₆
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5

Order Now

Specification Test Results

Assay (C₁₇H₂₀N₄O₆) 98.0 - 102.0 %
Specific Rotation [A]D -115 to -135
Loss On Drying max. 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR