Polyvinylpyrrolidone, M.W. 1 300 000 K-90 USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P1416-2.5KG P1416-12KG P1416-100GM P1416-500GM P1416-50KGBL
95035-258EA 2274.35 USD
95035-258 700008-860 95035-256 95035-260 89078-982 89078-984
Polyvinylpyrrolidone, M.W. 1 300 000 K-90 USP suitable for use as excipient
Polyvinylpyrrolidone

Povidone, K-90, USP is used as a binder in pharmaceutical tablets. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR