Potassium acetate ≥99.0%, crystalline powder USP

Supplier: Spectrum Chemical Mfg. Corp.

PO155-12KG PO155-500GM PO155-125GM PO155-2.5CS4 PO155-2.5KG PO155-500CS6
700004-360EA 1520.22 USD
700004-360 95035-076 95035-068 95035-070 95035-072 95035-074
Potassium acetate ≥99.0%, crystalline powder USP
Potassium acetate

POTASSIUM ACETATE, CRYSTALLINE POWDER, USP is, as its name suggests, the potassium salt of acetic acid and is used as a catalyst for producing polyurethanes. In the food industry it is used as a food additive, an acidity regulator and a preservative. It can also be found in medicine and biochemistry where it's employed to help treat diabetic ketoacidosis. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - DELIQUESCENT: Keep tightly closed.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CH₃COOK
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
pH of a 5% Solution @ 25°C 7.5 - 8.5
Loss On Drying max. 1.0 %
Heavy Metals max. 20 ppm
Sodium max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR