Polysorbate 80 FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

P1179-500MLG P1179-4LTGL P1179-200LTBL P1179-20LT
95035-054EA 190.16 USD
95035-054 95035-052 75811-808 75811-810
Polysorbate 80 FCC suitable for use as excipient
Polysorbate 80

Polysorbate 80, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kosher certified


Storage temperature - Keep tightly closed, protect from light.

Melting Pt: 100
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9

Order Now

Specification Test Results

Assay 65% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR