Polysorbate 20 NF

Supplier: Spectrum Chemical Mfg. Corp.

PO132-20LT PO132-200LTB PO132-4CS4 PO132-4LTGL PO132-500CS6 PO132-500MLG PO132-500MLG
700011-050EA 1214.49 USD
700011-050 700011-048 95035-036 95035-038 95035-040 700011-052 95035-042
Polysorbate 20 NF
Polysorbate 20

Polysorbate 20, NF is a hydrophilic, nonionic, polysorbate surfactant and a polyoxyethylene derivative of sorbitan monolaurate with relative non-toxicity. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₅₈H₁₁₄O₂₆
Density: 1.095
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR