Mineral oil, white (petroleum) light NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Matrix oil, Mineral oil, Paraffin oil

M1293-4CS4 M1293-1LT M1293-20LT M1293-4LTGL M1293-200LTB M1293-500MLG M1293-200LTB
95034-656CS 1439.8 USD
95034-656 95034-654 89078-900 95034-658 700000-452 700000-454 89078-902
Mineral oil, white (petroleum) light NF suitable for use as excipient
White mineral oil (petroleum)

Light Mineral Oil, NF is used as a vehicle and humectant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Boiling Pt: 300
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8042-47-5
EINECS: 232-455-8

Order Now

Specification Test Results

SPECIFIC GRAVITY @ 25°C 0.818-0.880
VISCOSITY @40.0 °C 3.0-34.4 mm²/s
ACIDITY TO PASS TEST
READILY CARBONIZABLE SUBSTANCES TO PASS TEST
POLYCYCLIC AROMATIC HYDROCARBONS TO PASS TEST
SOLID PARAFFIN TO PASS TEST
SULFUR COMPOUNDS TO PASS TEST
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS TO PASS TEST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR