Methyl-4-hydroxybenzoate ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

M1187-500GM M1187-25GM M1187-2.5KG M1187-12KG M1187-50KGBL
95034-510EA 205.1 USD
95034-510 95034-508 95034-506 75811-602 75811-604
Methyl-4-hydroxybenzoate ≥99.0% FCC
Methyl 4-hydroxybenzoate

Methylparaben, FCC is used as a food preservative and has the E number E218. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Melting Range 125 - 128 °C
Acidity To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR