Methyl cellulose, 15 cP (2% H₂O) USP

Supplier: Spectrum Chemical Mfg. Corp.

M1314-2.5KG M1314-500GM M1314-12KG M1314-45KGBL M1314-100GM
95034-466EA 1270.7 USD
95034-466 95034-468 700013-028 700013-030 95034-464
Methyl cellulose, 15 cP (2% H₂O) USP
Methyl cellulose

Methylcellulose, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Assay 26% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR