Manganese(II) sulfate monohydrate ≥98.0%, powder FCC, BP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

MA164-2.5KG MA164-500GM MA164-12KG MA164-50KGBL
95034-318EA 626.8 USD
95034-318 95034-320 75811-708 75811-710
Manganese(II) sulfate monohydrate ≥98.0%, powder FCC, BP
Manganese (II) sulphate monohydrate

Manganese Sulfate, Monohydrate, Powder, FCC, BP - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Health hazard

Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

Specification Test Results

Assay (MnSO₄[-H₂O) 98.0 - 102.0%
Assay (Ignited) 99.0 - 101.0%
Loss On Heating 10.0 - 12.0%
Appearance of Solution To Pass Test
Arsenic (As) max. 3 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR