Magnesium sulfate, anhydrous ≥99.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

M1354-2.5KG M1354-500GM M1354-50KGBL M1354-125GM M1354-12KG
95034-266EA 1031.69 USD
95034-266 95034-268 89078-866 95034-264 75811-646
Magnesium sulfate, anhydrous ≥99.0% USP
Magnesium sulphate

MAGNESIUM SULFATE, ANHYDROUS, USP is the raw material for a common magnesium-based, pharmaceutical preparation called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it's aNHydrous form, magnesium sulfate is used as a drying agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5 %
pH (In Solution 1 in 20) 5.0 - 9.2
Loss On Drying max. 2 %
Chloride (Cl) max. 0.014 %
Iron max. 20 Ug/G


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR