L-Phenylalanine ≥98.5% USP
Supplier: Spectrum Chemical Mfg. Corp.
Phenylalanine, USP is an amino acid, a building block of protein.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₉H₁₁NO₂ Boiling Pt: 313 °C (1013 hPa) Melting Pt: 173 °C Density: 0.754 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
Specification Test Results
Assay | 98.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...