L-Phenylalanine ≥98.5% USP

Supplier: Spectrum Chemical Mfg. Corp.

P1582-5KGBL P1582-100GM P1582-1KG P1582-25GM
700007-408EA 11512 USD
700007-408 95034-048 95034-050 95034-052
L-Phenylalanine ≥98.5% USP
L-α-Phenylalanine

Phenylalanine, USP is an amino acid, a building block of protein. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₉H₁₁NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 173 °C
Density: 0.754 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR