Hydroxypropyl methylcellulose (Hypromellose), 100 mPa.s USP Substitution Type 2208

Supplier: Spectrum Chemical Mfg. Corp.

HY123-45KGBL HY123-2.5KG HY123-12KG HY123-125GM HY123-500GM
700011-798EA 6186.75 USD
700011-798 95033-642 700011-796 95033-640 95033-644
Hydroxypropyl methylcellulose (Hypromellose), 100 mPa.s USP Substitution Type 2208
Hydroxypropyl methyl cellulose (Hypromellose)

Hypromellose, Substitution Type 2208, 100 mPa.s, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃H₈O₂·xCH₄O
Storage Temperature: Ambient
MDL Number: MFCD00131360
CAS Number: 9004-65-3

Order Now

Specification Test Results

Viscosity of a 2% Aqueous Solution @ 20D °C 80 - 120 mPa·s
Assay (Methoxy Groups) 19.0 - 24.0%
Assay (Hydroxypropoxy Groups) 4.0 - 12.0%
pH 5.0 - 8.0
Loss On Drying max. 5.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR