Folic acid ≥97.0%, powder USP

Supplier: Spectrum Chemical Mfg. Corp.

FO105-100GM FO105-1KG FO105-10GM FO105-25GM FO105-25KGBL FO105-5KGBL
95033-316EA 597.6 USD
95033-316 95033-320 95033-318 95033-322 700009-440 75811-208
Folic acid ≥97.0%, powder USP
Folic acid

 FOLIC ACID, POWDER, USP is a water soluble B vitamin. As a medication, folic acid is used to treat folic acid deficiency associated with ulcerative colitis, alcoholism, kidney dialysis, and liver disease and certain types of anemia. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₉H₁₉N₇O₆
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.0 - 102.0 %
Water Determination max.8.5 %
Residue On Ignition max. 0.3 %
Related Compounds max. 2.0 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR