Ethyl oleate NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

E1172-25GM E1172-100GM E1172-500GM E1172-5KGBL
95033-156EA 150.24 USD
95033-156 95033-154 95033-158 75811-060
Ethyl oleate NF
Ethyl oleate

Ethyl Oleate, NF is used as a solubilizing agent in pharmaceutical preparations. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₂₀H₃₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6

Order Now

Specification Test Results

Assay (As Ethyl (Z)-9-Octadecenoate) To Pass Test
Presence of Ester To Pass Test
Iodine Value 75 - 85
Peroxide Value max. 10.0
Water Determination max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR