EDTA disodium salt dihydrate ≥99% USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

ED150-2.5KG ED150-125GM ED150-2.5CS4 ED150-500CS6 ED150-45KGBL ED150-125CS6 ED150-12KG ED150-500GM
95033-068EA 1353.14 USD
95033-068 95033-064 95033-066 95033-070 89050-512 95033-062 89050-510 95033-072
EDTA disodium salt dihydrate ≥99% USP suitable for use as excipient
EDTA disodium salt dihydrate

Edetate Disodium, Dihydrate, USP is used for its heavy metal chelating properties. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR