Dimethyl sulfoxide ≥99.9% USP

Supplier: Spectrum Chemical Mfg. Corp.

D1258-500ML D1258-100ML
95032-948EA 1250.23 USD
95032-948 95032-944 95032-946
Dimethyl sulfoxide ≥99.9% USP
Dimethyl sulphoxide

Dimethyl Sulfoxide, USP is also known as DMSO. It is used topically to decrease pain and speed the healing of wounds, burns, and muscle and skeletal injuries. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: (CH₃)₂SO
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.100
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR