Calcium sulfate dihydrate ≥98.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

CA173-500GM CA173-2.5KG CA173-12KG
95032-348EA 174.55 USD
95032-348 95032-346 75810-478 75810-480
Calcium sulfate dihydrate ≥98.0% FCC
Calcium sulphate dihydrate

Calcium Sulfate, Dihydrate, FCC is widely used as pH regulator, hardness regulator, yeast agent, nutritional suppliment, texture and flavor enhancer in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CaSO₄·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 % min.
Loss On Drying 19.0 - 23.0 %
Fluoride max. 0.003 %
Lead (Pb) max. 2 mg/kg
Selenium (Se) max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR