Calcium chloride, anhydrous ≥93.0%, pellets FCC

Supplier: Spectrum Chemical Mfg. Corp.

C1977-2.5KG C1977-500GM C1977-12KG C1977-45KGBL
95032-238EA 476.98 USD
95032-238 95032-240 75810-864 75810-866
Calcium chloride, anhydrous ≥93.0%, pellets FCC

Calcium Chloride, Anhydrous, Pellets, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR