Calcium chloride, anhydrous ≥93%, granular FCC suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
Calcium Chloride, Anhydrous, Granular, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
CaCl₂ Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 772 °C Density: 2.174 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 REACH: 01-2119494219-28 Merck Index: 14,01659 |
Specification Test Results
Assay | 93% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...