Benzethonium chloride ≥97.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

BE129-25GMGL BE129-50KGBL BE129-12KGBL BE129-500GMG BE129-125GMG BE129-500GMG BE129-2.5KG
95031-972EA 117.64 USD
95031-972 89078-602 89078-600 700007-568 95031-970 95031-974 75810-690
Benzethonium chloride ≥97.0% USP
Benzethonium chloride

Benzethonium Chloride, USP exhibits a broad spectrum of microbiocidal activity against bacteria, fungi, mold and viruses. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₇H₄₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00011742
CAS Number: 121-54-0
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 103.0%
Melting Range 158° - 163°C
Loss On Drying max. 5.0%
Residue On Ignition max. 0.1%
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR