Propionic acid 99.5-100.5% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Propanoic acid

P1464-4CS4
95031-450CS 2248.9 USD
95031-450
Propionic acid 99.5-100.5% NF
Propionic acid

Propionic Acid, NF is used as a preservative in pharmaceutical formulations. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


Certifications: Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.993
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8.3,II
REACH: 01-2119486971-24
Merck Index: 13,07917

Order Now

Specification Test Results

Assay Min. 99.5 - 100.5%
Specific gravity at 25 °C 0.988 - 0.993
Distilling Range 138.5 - 142.5 °C
Nonvolatile Residue 0.0001
Elemental Impurities As Reported
Readily Oxidizable Substances To Pass Test
Aldehyde To Pass Test
Identification To Pass Test
Residual Solvents To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR