Zinc chloride ≥97.0%, granulated USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

ZI105-100GM ZI105-2.5KG ZI105-500GM ZI105-12KG ZI105-1.5KGBPL ZI105-50KGBL
95031-392EA 245.86 USD
95031-392 95031-394 95031-396 89079-050 75812-640 75812-642
Zinc chloride ≥97.0%, granulated USP
Zinc chloride

Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark
UN2331

Formula: ZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2331
ADR: 8,III

Order Now

Specification Test Results

Assay 97.0 - 100.5 %
Oxychloride To Pass Test
Sulfates (SO₄) max. 0.03 %
Alkalies and Alkaline Earths max. 1.0 %
Ammonium Salts To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR